Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LEP SIRNA Assays ›
          Gene Symbol:
          LEP
          Gene Name:
          leptin
          Gene Aliases:
          LEPD, OB, OBS
          Chromosome Location:
          Chr.7: 128241201 - 128257629 on Build GRCh38
          UniGene ID:
          Hs.194236
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS106015
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LEP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000230.2 NP_000221.1 3 222
          XM_005250340.4 XP_005250397.1 3 296
          GenBank mRNA AF008123.1 - 2 182
          AK313553.1 - 3 220
          BC060830.1 - 3 220 30333193
          BC069323.1 - 2 216 7262097
          BC069452.1 - 2 216 7262109
          BC069527.1 - 2 216 7262121
          D49487.1 - 2 179
          DA762132.1 - 3 222
          U18915.1 - 2 165
          U43653.1 - 2 221

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          ovulation from ovarian follicle
          response to hypoxia
          positive regulation of cytokine production
          placenta development
          regulation of endothelial cell proliferation
          response to dietary excess
          cardiac muscle hypertrophy
          glucose metabolic process
          regulation of gluconeogenesis
          energy reserve metabolic process
          glycerol biosynthetic process
          lipid metabolic process
          fatty acid beta-oxidation
          tyrosine phosphorylation of STAT protein
          female pregnancy
          circadian rhythm
          cholesterol metabolic process
          bile acid metabolic process
          regulation of blood pressure
          adult feeding behavior
          negative regulation of autophagy
          negative regulation of lipid storage
          positive regulation of phosphatidylinositol 3-kinase signaling
          response to activity
          sexual reproduction
          central nervous system neuron development
          insulin secretion
          T cell differentiation
          regulation of intestinal cholesterol absorption
          positive regulation of TOR signaling
          negative regulation of appetite
          prostaglandin secretion
          response to estradiol
          regulation of natural killer cell activation
          regulation of natural killer cell proliferation
          response to insulin
          response to vitamin E
          leptin-mediated signaling pathway
          positive regulation of luteinizing hormone secretion
          positive regulation of peroxisome proliferator activated receptor signaling pathway
          bone mineralization involved in bone maturation
          negative regulation of appetite by leptin-mediated signaling pathway
          positive regulation of T cell proliferation
          regulation of natural killer cell mediated cytotoxicity
          hormone metabolic process
          positive regulation of tyrosine phosphorylation of Stat3 protein
          glucose homeostasis
          eating behavior
          negative regulation of apoptotic process
          positive regulation of ion transport
          positive regulation of MAPK cascade
          cellular response to leptin stimulus
          response to ethanol
          positive regulation of myeloid cell differentiation
          regulation of angiogenesis
          negative regulation of vasoconstriction
          negative regulation of glucose import
          positive regulation of JAK-STAT cascade
          positive regulation of insulin receptor signaling pathway
          regulation of bone remodeling
          positive regulation of follicle-stimulating hormone secretion
          positive regulation of developmental growth
          regulation of insulin secretion
          regulation of steroid biosynthetic process
          intestinal absorption
          leukocyte tethering or rolling
          regulation of nitric-oxide synthase activity
          regulation of cell cycle
          positive regulation of protein kinase B signaling
          regulation of lipoprotein lipid oxidation
          adipose tissue development
          negative regulation of cartilage development
          negative regulation of glucagon secretion
          cellular response to L-ascorbic acid
          cellular response to retinoic acid
          interleukin-6 secretion
          interleukin-8 secretion
          regulation of brown fat cell differentiation
          bone growth
          regulation of cytokine production involved in inflammatory response
          positive regulation of p38MAPK cascade
          positive regulation of fat cell apoptotic process
          activation of protein kinase C activity
          positive regulation of STAT protein import into nucleus
          positive regulation of reactive oxygen species metabolic process
          negative regulation of glutamine transport
          positive regulation of hepatic stellate cell activation
          hormone activity
          growth factor activity
          peptide hormone receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0