Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABLIM1 SIRNA Assays ›
Gene Symbol:
ABLIM1
Gene Name:
actin binding LIM protein 1
Gene Aliases:
ABLIM, LIMAB1, LIMATIN, abLIM-1
Chromosome Location:
Chr.10: 114431110 - 114821643 on Build GRCh38
UniGene ID:
Hs.438236
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS106064
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABLIM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003407.1
NP_001003407.1
17
2010
NM_001003408.1
NP_001003408.1
18
2094
NM_001322882.1
NP_001309811.1
18
2100
NM_001322883.1
NP_001309812.1
16
1905
NM_001322884.1
NP_001309813.1
16
1710
NM_001322885.1
NP_001309814.1
16
1704
NM_001322886.1
NP_001309815.1
16
1668
NM_001322887.1
NP_001309816.1
15
1764
NM_001322888.1
NP_001309817.1
12
1240
NM_001322889.1
NP_001309818.1
12
1225
NM_001322890.1
NP_001309819.1
11
1162
NM_001322891.1
NP_001309820.1
11
1183
NM_001322892.1
NP_001309821.1
11
1156
NM_001322893.1
NP_001309822.1
11
1177
NM_001322894.1
NP_001309823.1
11
1141
NM_001322895.1
NP_001309824.1
11
1120
NM_001322896.1
NP_001309825.1
10
1078
NM_001322897.1
NP_001309826.1
10
1042
NM_001322898.1
NP_001309827.1
10
1015
NM_001322899.1
NP_001309828.1
10
1036
NM_001322900.1
NP_001309829.1
9
937
NM_002313.5
NP_002304.3
17
1987
NM_006720.3
NP_006711.3
12
1240
XM_005269824.2
XP_005269881.1
16
2605
XM_005269826.1
XP_005269883.1
14
1476
XM_005269827.4
XP_005269884.1
13
2106
XM_006717837.1
XP_006717900.1
19
2568
XM_006717846.3
XP_006717909.1
14
2211
XM_006717847.1
XP_006717910.1
13
1305
XM_011539801.1
XP_011538103.1
19
1972
XM_011539802.1
XP_011538104.1
19
1885
XM_011539803.1
XP_011538105.1
19
1864
XM_011539805.1
XP_011538107.1
19
2225
XM_017016242.1
XP_016871731.1
21
2068
XM_017016243.1
XP_016871732.1
20
1954
XM_017016244.1
XP_016871733.1
20
1948
XM_017016245.1
XP_016871734.1
19
2058
XM_017016246.1
XP_016871735.1
19
1849
XM_017016247.1
XP_016871736.1
16
1623
XM_017016248.1
XP_016871737.1
16
1623
XM_017016249.1
XP_016871738.1
19
1807
XM_017016250.1
XP_016871739.1
18
1974
XM_017016251.1
XP_016871740.1
18
1765
XM_017016252.1
XP_016871741.1
18
1759
XM_017016253.1
XP_016871742.1
18
1723
XM_017016254.1
XP_016871743.1
18
1702
XM_017016255.1
XP_016871744.1
17
1854
XM_017016256.1
XP_016871745.1
16
2647
XM_017016258.1
XP_016871747.1
12
1986
XM_017016259.1
XP_016871748.1
12
1185
GenBank mRNA
AF005654.1
-
17
2008
AK058007.1
-
7
629
AK093471.1
-
17
1509
AK098277.1
-
12
1245
AK122891.1
-
18
2450
AK297344.1
-
16
1943
AK298709.1
-
10
919
BC002448.2
-
10
982
3347450
CR749819.1
-
11
1152
D31883.2
-
17
1812
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
visual perception
organ morphogenesis
lamellipodium assembly
cilium assembly
actin binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-49ts5:80/100.66.72.225:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0