Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LMO7 SIRNA Assays ›
Gene Symbol:
LMO7
Gene Name:
LIM domain 7
Gene Aliases:
FBX20, FBXO20, LOMP
Chromosome Location:
Chr.13: 75620434 - 75859870 on Build GRCh38
UniGene ID:
Hs.207631
Species:
Human
siRNA ID
HSS106110
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LMO7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306080.1
NP_001293009.1
24
4128
NM_005358.5
NP_005349.3
23
4172
NM_015842.2
NP_056667.2
21
3819
XM_011535075.2
XP_011533377.1
25
4016
XM_011535078.1
XP_011533380.1
24
3974
XM_011535080.2
XP_011533382.1
25
3935
XM_011535082.1
XP_011533384.1
24
3893
XM_011535086.1
XP_011533388.1
23
3689
XM_011535094.1
XP_011533396.1
18
2597
XM_011535095.1
XP_011533397.1
21
2768
XM_017020574.1
XP_016876063.1
25
4016
XM_017020575.1
XP_016876064.1
24
3974
XM_017020576.1
XP_016876065.1
24
3959
XM_017020577.1
XP_016876066.1
25
3935
XM_017020578.1
XP_016876067.1
23
3917
XM_017020579.1
XP_016876068.1
24
3893
XM_017020580.1
XP_016876069.1
24
3812
XM_017020581.1
XP_016876070.1
24
3938
XM_017020582.1
XP_016876071.1
24
3731
XM_017020583.1
XP_016876072.1
24
3731
XM_017020584.1
XP_016876073.1
23
3734
XM_017020585.1
XP_016876074.1
23
3530
XM_017020586.1
XP_016876075.1
23
3449
XM_017020587.1
XP_016876076.1
22
3407
XM_017020588.1
XP_016876077.1
22
3488
XM_017020589.1
XP_016876078.1
22
3611
XM_017020590.1
XP_016876079.1
22
3407
XM_017020591.1
XP_016876080.1
24
4099
XM_017020592.1
XP_016876081.1
24
3982
XM_017020593.1
XP_016876082.1
23
3895
XM_017020594.1
XP_016876083.1
23
3778
XM_017020595.1
XP_016876084.1
22
3772
XM_017020596.1
XP_016876085.1
22
2810
XM_017020597.1
XP_016876086.1
21
2780
XM_017020598.1
XP_016876087.1
22
3772
XM_017020599.1
XP_016876088.1
22
3613
XM_017020600.1
XP_016876089.1
22
2729
XM_017020601.1
XP_016876090.1
21
3490
XM_017020602.1
XP_016876091.1
21
3373
XM_017020603.1
XP_016876092.1
21
3490
XM_017020604.1
XP_016876093.1
21
2893
GenBank mRNA
AB020665.2
-
21
3749
AF144237.1
-
12
1632
AF330045.1
-
23
4171
AK055465.1
-
16
2784
AK074265.1
-
8
621
AK092052.1
-
22
3091
AK304051.1
-
19
2710
FJ711162.1
-
24
3758
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination
regulation of signaling
regulation of cell adhesion
ubiquitin-protein transferase activity
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-gzxth:80/10.239.166.29:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0