Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322468.1 | NP_001309397.1 | 20494 | |||
NM_004532.5 | NP_004523.3 | 12 | 1768 | |||
NM_018406.6 | NP_060876.5 | 14476 | ||||
NM_138297.4 | NP_612154.2 | 11 | 1615 | |||
GenBank mRNA | AJ010901.1 | - | 12 | 1552 | ||
AJ242541.1 | - | 13 | 1777 | |||
AJ242542.1 | - | 11 | 1477 | |||
AJ242543.1 | - | 12 | 1476 | |||
AJ242544.1 | - | 12 | 1448 | |||
AJ242545.1 | - | 11 | 1387 | |||
AJ242546.1 | - | 10 | 1122 | |||
AJ242547.1 | - | 12 | 1552 | |||
AJ242549.1 | - | 12 | 1652 | |||
AJ242550.1 | - | 11 | 1499 | |||
AJ276359.1 | - | 14 | 4662 | |||
AJ277412.1 | - | 13 | 4506 | |||
AJ277505.1 | - | 12 | 4403 | |||
AJ400633.1 | - | 14 | 4676 | |||
AJ400849.1 | - | 14 | 4586 | |||
AJ400850.1 | - | 14 | 4586 | |||
AJ400851.1 | - | 14 | 4676 | |||
AJ400852.1 | - | 13 | 4430 | |||
AJ400853.1 | - | 13 | 4402 | |||
AJ400854.1 | - | 13 | 4507 | |||
AJ400855.1 | - | 13 | 4478 | |||
AJ400856.1 | - | 14 | 4633 | |||
AJ400857.1 | - | 14 | 4662 | |||
AJ400858.1 | - | 14 | 4662 |