Search
Search

MYH3| Gene Symbol: MYH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002470.3 | NP_002461.2 | 10, 11 | 984 | ||
| XM_011523870.2 | XP_011522172.1 | 10, 11 | 2351 | |||
| XM_011523871.2 | XP_011522173.1 | 10, 11 | 984 | |||
| GenBank mRNA | X13988.1 | - | 9, 10 | 980 | ||