Search
Search

NDUFS3| Gene Symbol: NDUFS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004551.2 | NP_004542.1 | 3, 4 | 301 | ||
| GenBank mRNA | AF067139.1 | - | 3, 4 | 231 | ||
| AF100743.1 | - | 3, 4 | 228 | |||
| AK223255.1 | - | 3, 4 | 233 | |||
| AK294167.1 | - | 3, 4 | 226 | |||
| AK313802.1 | - | 3, 4 | 233 | |||
| BC000617.2 | - | 3, 4 | 239 | 3346129 | ||
| DA893277.1 | - | 3, 4 | 301 | |||