Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199397.1 | NP_001186326.1 | 31 | 3784 | ||
NM_001199398.1 | NP_001186327.1 | 30 | 3652 | |||
NM_001199399.1 | NP_001186328.1 | 28 | 3493 | |||
NM_001199400.1 | NP_001186329.1 | 28 | 3646 | |||
NM_012224.2 | NP_036356.1 | 29 | 3778 | |||
XM_006714228.1 | XP_006714291.1 | 30 | 3609 | |||
XM_011532003.1 | XP_011530305.1 | 30 | 3702 | |||
XM_011532004.1 | XP_011530306.1 | 29 | 3570 | |||
XM_017008249.1 | XP_016863738.1 | 30 | 3688 | |||
XM_017008250.1 | XP_016863739.1 | 29 | 3806 | |||
XM_017008251.1 | XP_016863740.1 | 29 | 3604 | |||
XM_017008252.1 | XP_016863741.1 | 27 | 3624 | |||
XM_017008253.1 | XP_016863742.1 | 30 | 3634 | |||
XM_017008254.1 | XP_016863743.1 | 23 | 2744 | |||
GenBank mRNA | AB067488.1 | - | 29 | 3614 | ||
AF155113.1 | - | 13 | 1662 | |||
AK025658.1 | - | 4 | 532 | |||
AL050385.1 | - | 30 | 3608 | |||
BC037790.2 | - | 3 | 444 | |||
BC114491.1 | - | 28 | 3253 | 40082305 | ||
CR933642.1 | - | 31 | 3742 | |||
CR936672.1 | - | 28 | 3604 |