Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TONSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013432.4 | NP_038460.4 | 17 | 2676 | ||
XM_011517048.2 | XP_011515350.1 | 10 | 1775 | |||
XM_011517049.2 | XP_011515351.1 | 10 | 1736 | |||
XM_011517050.2 | XP_011515352.1 | 17 | 2735 | |||
GenBank mRNA | AB209126.1 | - | 9 | 3636 | ||
BC008782.2 | - | 16 | 2648 | 3628374 |