Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002507.3 | NP_002498.1 | 6 | 1240 | ||
GenBank mRNA | AK303278.1 | - | 6 | 1308 | ||
AK313654.1 | - | 6 | 1240 | |||
BC050309.1 | - | 6 | 1240 | 5263715 | ||
M14764.1 | - | 6 | 1228 |