Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000620.4 | NP_000611.1 | 10 | 2433 | ||
NM_001204213.1 | NP_001191142.1 | 9 | 1025 | |||
NM_001204214.1 | NP_001191143.1 | 9 | 1020 | |||
NM_001204218.1 | NP_001191147.1 | 10 | 2433 | |||
XM_011538398.2 | XP_011536700.1 | 11 | 2304 | |||
XM_017019345.1 | XP_016874834.1 | 10 | 2440 | |||
XM_017019346.1 | XP_016874835.1 | 10 | 2254 | |||
XM_017019347.1 | XP_016874836.1 | 10 | 2343 | |||
GenBank mRNA | AK294435.1 | - | 10 | 2237 | ||
AK307481.1 | - | 10 | 2433 | |||
D16408.1 | - | 9 | 1722 | |||
EU753241.1 | - | 10 | 2407 | |||
L02881.1 | - | 9 | 2149 | |||
U17327.1 | - | 10 | 2407 | |||
U31466.1 | - | 9 | 1729 |