siRNA Details
Gene Symbol: NOTCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017617.4 | NP_060087.3 | 34 | 6912 | ||
XM_011518717.2 | XP_011517019.2 | 31 | 6204 | |||
GenBank mRNA | AB209873.1 | - | 26 | 5449 | ||
AF308602.1 | - | 34 | 6915 | |||
BC013208.1 | - | 4 | 1097 | |||
M73980.1 | - | 34 | 6915 |

More Information
Panther Classification:
Molecular Function -




