Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032409.2 | NP_001027581.1 | 2 | 582 | ||
NM_001320151.1 | NP_001307080.1 | 2 | 582 | |||
NM_002534.3 | NP_002525.2 | 2 | 582 | |||
NM_016816.3 | NP_058132.2 | 2 | 582 | |||
XM_006719434.1 | XP_006719497.1 | 2 | 409 | |||
XM_011538413.1 | XP_011536715.1 | 2 | 409 | |||
XM_017019361.1 | XP_016874850.1 | 2 | 409 | |||
XM_017019362.1 | XP_016874851.1 | 2 | 281 | |||
GenBank mRNA | AJ629455.1 | - | 2 | 352 | ||
AK223006.1 | - | 2 | 341 | |||
AK225384.1 | - | 2 | 382 | |||
AK291003.1 | - | 2 | 386 | |||
AK300346.1 | - | 2 | 307 | |||
AK301498.1 | - | 2 | 341 | |||
AY730627.1 | - | 2 | 328 | |||
AY730628.1 | - | 2 | 328 | |||
BC000562.2 | - | 2 | 389 | 3162421 | ||
BC061587.1 | - | 2 | 350 | 6502031 | ||
BT006785.1 | - | 2 | 319 | |||
D00068.1 | - | 2 | 382 | |||
X02874.1 | - | 2 | 352 | |||
X04371.1 | - | 2 | 412 |