Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other P2RX7 SIRNA Assays ›
          Gene Symbol:
          P2RX7
          Gene Name:
          purinergic receptor P2X 7
          Gene Aliases:
          P2X7
          Chromosome Location:
          Chr.12: 121132819 - 121189478 on Build GRCh38
          UniGene ID:
          Hs.729169
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS107529
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: P2RX7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002562.5 NP_002553.3 2, 3 448
          NR_033948.1 3, 4 587
          NR_033949.1 3, 4 587
          NR_033950.1 2, 3 448
          NR_033951.1 2, 3 448
          NR_033952.1 2, 3 448
          NR_033953.1 1, 2 288
          NR_033954.1 2, 3 448
          NR_033955.1 2, 3 448
          NR_033956.1 2, 3 448
          XM_011538419.2 XP_011536721.1 1, 2 288
          XM_011538420.2 XP_011536722.1 1, 2 92
          XM_017019364.1 XP_016874853.1 2, 3 448
          XM_017019365.1 XP_016874854.1 1, 2 176
          XM_017019366.1 XP_016874855.1 2, 3 389
          XM_017019367.1 XP_016874856.1 1, 2 227
          GenBank mRNA AB209709.1 - 2, 3 369
          AK090866.1 - 1, 2 165
          AK225163.1 - 2, 3 334
          AK290405.1 - 2, 3 396
          AK294126.1 - 2, 3 399
          AK296432.1 - 2, 3 396
          AY847298.1 - 2, 3 305
          AY847299.1 - 2, 3 305
          AY847300.1 - 2, 3 305
          AY847301.1 - 2, 3 305
          AY847302.1 - 2, 3 305
          AY847303.1 - 3, 4 444
          AY847304.1 - 3, 4 444
          BC007679.2 - 2, 3 346 3628076
          BC011913.2 - 2, 3 374 4298811
          BC121157.1 - 2, 3 374
          BC121158.1 - 2, 3 374
          BI598073.1 - 2, 3 438
          DA274887.1 - 3, 4 473
          DA302256.1 - 1, 2 288
          DQ399293.1 - 2, 3 305
          GQ180122.1 - 2, 3 314
          Y09561.1 - 2, 3 331

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          cell morphogenesis
          phagolysosome assembly
          positive regulation of T cell mediated cytotoxicity
          regulation of sodium ion transport
          protein phosphorylation
          membrane protein ectodomain proteolysis
          phospholipid transfer to membrane
          membrane budding
          inflammatory response
          mitochondrion organization
          cell surface receptor signaling pathway
          blood coagulation
          response to mechanical stimulus
          response to zinc ion
          positive regulation of calcium ion transport into cytosol
          positive regulation of gene expression
          positive regulation of glutamate secretion
          positive regulation of gamma-aminobutyric acid secretion
          synaptic vesicle exocytosis
          protein processing
          phospholipid scrambling
          sensory perception of pain
          cytolysis
          positive regulation of bone mineralization
          cellular response to extracellular stimulus
          bleb assembly
          positive regulation of prostaglandin secretion
          response to lipopolysaccharide
          positive regulation of interleukin-6 production
          collagen metabolic process
          response to ATP
          response to fluid shear stress
          positive regulation of ion transmembrane transport
          purinergic nucleotide receptor signaling pathway
          T cell proliferation
          response to drug
          T cell homeostasis
          NAD transport
          negative regulation of MAPK cascade
          multicellular organismal protein catabolic process
          phospholipid translocation
          negative regulation of bone resorption
          negative regulation of cell volume
          positive regulation of glycolytic process
          positive regulation of cytolysis
          ceramide biosynthetic process
          pore complex assembly
          skeletal system morphogenesis
          homeostasis of number of cells within a tissue
          positive regulation of interleukin-1 alpha secretion
          positive regulation of interleukin-1 beta secretion
          defense response to Gram-positive bacterium
          release of sequestered calcium ion into cytosol
          protein oligomerization
          positive regulation of cytoskeleton organization
          response to calcium ion
          response to electrical stimulus
          regulation of killing of cells of other organism
          membrane depolarization
          positive regulation of mitochondrial depolarization
          positive regulation of lymphocyte apoptotic process
          cellular response to dsRNA
          reactive oxygen species metabolic process
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          positive regulation of bleb assembly
          lipopolysaccharide binding
          purinergic nucleotide receptor activity
          extracellular ATP-gated cation channel activity
          receptor binding
          protein binding
          ATP binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-jz86s:80/100.66.72.197:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0