Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAK3 SIRNA Assays ›
Gene Symbol:
PAK3
Gene Name:
p21 (RAC1) activated kinase 3
Gene Aliases:
ARA, MRX30, MRX47, OPHN3, PAK-3, PAK3beta, bPAK, beta-PAK
Chromosome Location:
Chr.X: 110944241 - 111227361 on Build GRCh38
UniGene ID:
Hs.593599
Species:
Human
siRNA ID
HSS107582
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128166.2
NP_001121638.1
13
1664
NM_001128167.2
NP_001121639.1
12
1591
NM_001128168.2
NP_001121640.1
17
1913
NM_001128172.2
NP_001121644.1
12
1266
NM_001128173.2
NP_001121645.1
16
1850
NM_001324325.1
NP_001311254.1
13
1742
NM_001324326.1
NP_001311255.1
13
1587
NM_001324327.1
NP_001311256.1
15
1699
NM_001324328.1
NP_001311257.1
14
1632
NM_001324329.1
NP_001311258.1
16
1847
NM_001324330.1
NP_001311259.1
13
1739
NM_001324331.1
NP_001311260.1
14
1654
NM_001324332.1
NP_001311261.1
15
1805
NM_001324333.1
NP_001311262.1
15
1780
NM_001324334.1
NP_001311263.1
14
1735
NM_002578.4
NP_002569.1
15
1802
NR_136747.1
14
1654
NR_136748.1
12
1351
XM_005262132.2
XP_005262189.1
16
1842
XM_005262133.3
XP_005262190.1
15
1797
XM_006724654.2
XP_006724717.1
15
1689
XM_006724655.2
XP_006724718.1
14
1632
XM_011530962.1
XP_011529264.1
16
1840
XM_011530967.2
XP_011529269.1
15
1780
XM_011530968.2
XP_011529270.1
16
1856
XM_011530969.1
XP_011529271.1
14
1701
XM_017029557.1
XP_016885046.1
14
1701
XM_017029558.1
XP_016885047.1
14
1644
XM_017029559.1
XP_016885048.1
13
1656
XM_017029560.1
XP_016885049.1
13
1587
XM_017029561.1
XP_016885050.1
15
1804
XM_017029562.1
XP_016885051.1
16
1842
XM_017029563.1
XP_016885052.1
17
1905
GenBank mRNA
AB102659.1
-
12
1221
AF068864.1
-
11
1176
AF155651.1
-
15
1567
AK128022.1
-
14
1909
AK290504.1
-
12
1591
AK315284.1
-
13
1508
AM943850.1
-
12
1221
AM943851.1
-
12
1239
AM943852.1
-
13
1284
BC117353.1
-
12
1270
BC117355.1
-
12
1270
BC143544.1
-
12
1270
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
mitotic cell cycle
stimulatory C-type lectin receptor signaling pathway
axonogenesis
dendrite development
regulation of actin filament polymerization
T cell costimulation
positive regulation of neuron apoptotic process
ephrin receptor signaling pathway
synapse organization
T cell receptor signaling pathway
dendritic spine morphogenesis
positive regulation of dendritic spine morphogenesis
cellular response to organic cyclic compound
positive regulation of DNA biosynthetic process
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase activity
protein binding
ATP binding
Rho GTPase binding
SH3 domain binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-89mxr:80/10.239.163.131:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0