Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAWR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002583.2 | NP_002574.2 | 5 | 1042 | ||
XM_006719435.2 | XP_006719498.1 | 5 | 1018 | |||
XM_017019377.1 | XP_016874866.1 | 5 | 922 | |||
GenBank mRNA | BC007018.1 | - | 4 | 888 | 3828412 | |
CR536549.1 | - | 4 | 756 | |||
U63809.1 | - | 5 | 1000 |