Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCM1 SIRNA Assays ›
Gene Symbol:
PCM1
Gene Name:
pericentriolar material 1
Gene Aliases:
PTC4, RET/PCM-1
Chromosome Location:
Chr.8: 17922763 - 18029948 on Build GRCh38
UniGene ID:
Hs.491148
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS107660
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001315507.1
NP_001302436.1
34, 35
5916
NM_006197.3
NP_006188.3
34, 35
5940
XM_006716336.3
XP_006716399.1
37, 38
6391
XM_006716340.3
XP_006716403.1
37, 38
6379
XM_006716349.3
XP_006716412.1
33, 34
5851
XM_011544528.2
XP_011542830.1
37, 38
6412
XM_011544529.2
XP_011542831.1
39, 40
6718
XM_011544530.2
XP_011542832.1
38, 39
6534
XM_011544532.2
XP_011542834.1
37, 38
6409
XM_011544533.2
XP_011542835.1
37, 38
6400
XM_011544534.2
XP_011542836.1
37, 38
6388
XM_011544535.2
XP_011542837.1
37, 38
6376
XM_011544540.2
XP_011542842.1
37, 38
6340
XM_017013472.1
XP_016868961.1
38, 39
6528
XM_017013473.1
XP_016868962.1
38, 39
6524
XM_017013474.1
XP_016868963.1
35, 36
6142
XM_017013475.1
XP_016868964.1
37, 38
6266
XM_017013476.1
XP_016868965.1
34, 35
5850
XM_017013477.1
XP_016868966.1
37, 38
6431
XM_017013478.1
XP_016868967.1
37, 38
6409
XM_017013479.1
XP_016868968.1
37, 38
6410
XM_017013480.1
XP_016868969.1
35, 36
6082
XM_017013481.1
XP_016868970.1
37, 38
6385
XM_017013482.1
XP_016868971.1
34, 35
6046
XM_017013483.1
XP_016868972.1
36, 37
6315
XM_017013484.1
XP_016868973.1
33, 34
5741
XM_017013485.1
XP_016868974.1
36, 37
6295
XM_017013486.1
XP_016868975.1
34, 35
5977
XM_017013487.1
XP_016868976.1
34, 35
5968
XM_017013488.1
XP_016868977.1
34, 35
5965
XM_017013489.1
XP_016868978.1
36, 37
6268
XM_017013490.1
XP_016868979.1
36, 37
6244
XM_017013491.1
XP_016868980.1
33, 34
5863
XM_017013492.1
XP_016868981.1
33, 34
5860
XM_017013493.1
XP_016868982.1
35, 36
6163
XM_017013494.1
XP_016868983.1
33, 34
5848
XM_017013495.1
XP_016868984.1
35, 36
5988
XM_017013496.1
XP_016868985.1
32, 33
5571
XM_017013497.1
XP_016868986.1
33, 34
5836
XM_017013498.1
XP_016868987.1
33, 34
5827
XM_017013499.1
XP_016868988.1
35, 36
5967
XM_017013500.1
XP_016868989.1
33, 34
5824
XM_017013501.1
XP_016868990.1
35, 36
6126
XM_017013502.1
XP_016868991.1
32, 33
5547
GenBank mRNA
AF068293.1
-
3, 4
441
AK091406.1
-
11, 12
1625
AK295856.1
-
12, 13
1926
AK301727.1
-
11, 12
1432
AK302378.1
-
13, 14
1737
AX785390.1
-
32, 33
5518
BC140946.1
-
34, 35
5745
9056758
BG898673.1
-
4, 5
503
L27841.1
-
34, 35
5927
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
neuron migration
interkinetic nuclear migration
cytoplasmic microtubule organization
microtubule anchoring
microtubule anchoring at centrosome
nonmotile primary cilium assembly
intraciliary transport involved in cilium morphogenesis
cilium assembly
negative regulation of neurogenesis
centrosome organization
protein localization to centrosome
positive regulation of intracellular protein transport
neuronal stem cell population maintenance
protein binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-gbx7x:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0