Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PKD1 SIRNA Assays ›
          Gene Symbol:
          PKD1
          Gene Name:
          polycystin 1, transient receptor potential channel interacting
          Gene Aliases:
          PBP, Pc-1, TRPP1
          Chromosome Location:
          Chr.16: 2088708 - 2135898 on Build GRCh38
          UniGene ID:
          Hs.75813
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS108041
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PKD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000296.3 NP_000287.3 37 11201
          NM_001009944.2 NP_001009944.2 37 11204
          XM_005255370.3 XP_005255427.1 27 8791
          XM_011522528.2 XP_011520830.1 38 11229
          XM_011522529.2 XP_011520831.1 38 11227
          XM_011522530.2 XP_011520832.1 38 11034
          XM_011522532.2 XP_011520834.1 37 10962
          XM_011522534.2 XP_011520836.1 38 11019
          XM_011522535.2 XP_011520837.1 29 9251
          XM_011522537.2 XP_011520839.1 27 8162
          XM_017023285.1 XP_016878774.1 28 8806
          GenBank mRNA AB209025.1 - 24 5558
          AB209675.1 - 23 6510
          L33243.1 - 37 11203
          U24497.1 - 37 11130

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          cartilage condensation
          in utero embryonic development
          kidney development
          liver development
          embryonic placenta development
          protein export from nucleus
          nitrogen compound metabolic process
          cell cycle arrest
          homophilic cell adhesion via plasma membrane adhesion molecules
          cell-matrix adhesion
          calcium-independent cell-matrix adhesion
          positive regulation of cytosolic calcium ion concentration
          JAK-STAT cascade
          heart development
          anatomical structure morphogenesis
          single organismal cell-cell adhesion
          peptidyl-serine phosphorylation
          spinal cord development
          neural tube development
          establishment of cell polarity
          regulation of cell adhesion
          positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
          positive regulation of protein binding
          response to fluid shear stress
          lymph vessel morphogenesis
          cytoplasmic sequestering of transcription factor
          skin development
          positive regulation of transcription from RNA polymerase II promoter
          digestive tract development
          branching morphogenesis of an epithelial tube
          genitalia development
          detection of mechanical stimulus
          cartilage development
          regulation of mitotic spindle organization
          lung epithelium development
          placenta blood vessel development
          regulation of proteasomal protein catabolic process
          calcium ion transmembrane transport
          mesonephric tubule development
          mesonephric duct development
          metanephric collecting duct development
          metanephric ascending thin limb development
          metanephric proximal tubule development
          metanephric distal tubule morphogenesis
          calcium channel activity
          protein binding
          protein kinase binding
          protein domain specific binding
          carbohydrate binding
          ion channel binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0