Search
Search

PPP2R5B| Gene Symbol: PPP2R5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006244.3 | NP_006235.1 | 8 | 1489 | ||
| XM_011545132.2 | XP_011543434.1 | 9 | 922 | |||
| XM_011545133.1 | XP_011543435.1 | 10 | 1042 | |||
| XM_011545134.2 | XP_011543436.1 | 8 | 2328 | |||
| GenBank mRNA | AK092730.1 | - | 8 | 1473 | ||
| BC045619.1 | - | 8 | 1483 | 5294756 | ||
| L42374.1 | - | 8 | 1192 | |||