Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PROX1 SIRNA Assays ›
          Gene Symbol:
          PROX1
          Gene Name:
          prospero homeobox 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 213983239 - 214041510 on Build GRCh38
          UniGene ID:
          Hs.741808
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS108597
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PROX1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270616.1 NP_001257545.1 2 1062
          NM_002763.4 NP_002754.2 2 735
          XM_005273194.1 XP_005273251.1 2 1061
          XM_005273195.4 XP_005273252.1 2 1090
          XM_011509771.1 XP_011508073.1 3 1169
          XM_011509772.2 XP_011508074.1 2 958
          XM_011509773.2 XP_011508075.1 2 599
          XM_017001832.1 XP_016857321.1 2 919
          XM_017001833.1 XP_016857322.1 2 919
          GenBank mRNA BC024201.2 - 2 719 3532312
          U44060.1 - 2 1053

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell fate determination
          kidney development
          liver development
          positive regulation of endothelial cell proliferation
          lymphangiogenesis
          lens development in camera-type eye
          hepatocyte cell migration
          transcription, DNA-templated
          brain development
          circadian rhythm
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          regulation of gene expression
          positive regulation of endothelial cell migration
          dorsal spinal cord development
          dentate gyrus development
          cerebellar granule cell differentiation
          neural tube development
          neuron differentiation
          skeletal muscle thin filament assembly
          lung development
          olfactory placode formation
          pancreas development
          response to nutrient levels
          regulation of circadian rhythm
          otic placode formation
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of viral genome replication
          positive regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          optic placode formation involved in camera-type eye formation
          venous blood vessel morphogenesis
          ventricular cardiac myofibril assembly
          cardiac muscle cell differentiation
          atrial cardiac muscle tissue morphogenesis
          ventricular cardiac muscle tissue morphogenesis
          retina morphogenesis in camera-type eye
          embryonic retina morphogenesis in camera-type eye
          endocardium formation
          positive regulation of sarcomere organization
          ventricular septum morphogenesis
          aorta smooth muscle tissue morphogenesis
          positive regulation of heart growth
          lymphatic endothelial cell differentiation
          regulation of transcription involved in lymphatic endothelial cell fate commitment
          branching involved in pancreas morphogenesis
          lens fiber cell morphogenesis
          hepatocyte differentiation
          negative regulation of bile acid biosynthetic process
          hepatocyte proliferation
          acinar cell differentiation
          neuronal stem cell population maintenance
          positive regulation of cell cycle checkpoint
          positive regulation of neural precursor cell proliferation
          positive regulation of forebrain neuron differentiation
          core promoter sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          transcription corepressor activity
          protein binding
          ligand-dependent nuclear receptor binding
          transcription regulatory region DNA binding
          DBD domain binding
          LBD domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0