Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRPSAP2 SIRNA Assays ›
Gene Symbol:
PRPSAP2
Gene Name:
phosphoribosyl pyrophosphate synthetase associated protein 2
Gene Aliases:
PAP41
Chromosome Location:
Chr.17: 18856299 - 18931287 on Build GRCh38
UniGene ID:
Hs.632236
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS108609
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRPSAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243936.1
NP_001230865.1
3
339
NM_001243940.1
NP_001230869.1
2
208
NM_001243941.1
NP_001230870.1
2
306
NM_001243942.1
NP_001230871.1
3
339
NM_002767.3
NP_002758.1
3
402
XM_005256725.2
XP_005256782.1
2
256
XM_005256726.1
XP_005256783.1
2
290
XM_005256729.4
XP_005256786.1
2
205
XM_017024864.1
XP_016880353.1
3
266
XM_017024865.1
XP_016880354.1
4
475
XM_017024866.1
XP_016880355.1
4
409
XM_017024867.1
XP_016880356.1
2
243
XM_017024868.1
XP_016880357.1
3
1095
XM_017024869.1
XP_016880358.1
3
374
XM_017024870.1
XP_016880359.1
3
289
XM_017024871.1
XP_016880360.1
3
352
XM_017024872.1
XP_016880361.1
4
488
XM_017024873.1
XP_016880362.1
3
337
XM_017024874.1
XP_016880363.1
3
409
XM_017024875.1
XP_016880364.1
3
282
XM_017024876.1
XP_016880365.1
2
260
GenBank mRNA
AB007851.1
-
2
167
AK303911.1
-
3
285
AK304544.1
-
2
191
AK313087.1
-
3
339
AL535304.3
-
3
299
BC101670.1
-
2
128
8069176
BC101672.1
-
2
128
8069178
BC106050.1
-
3
318
6176727
BC143475.1
-
2
128
9051983
BX648850.1
-
3
288
CR457082.1
-
1
80
DA257655.1
-
2
290
DC424578.1
-
3
402
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
nucleobase-containing compound metabolic process
nucleoside metabolic process
nucleotide biosynthetic process
negative regulation of catalytic activity
bone development
magnesium ion binding
ribose phosphate diphosphokinase activity
enzyme inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-x54js:80/100.66.79.231:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0