Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RELN SIRNA Assays ›
Gene Symbol:
RELN
Gene Name:
reelin
Gene Aliases:
ETL7, LIS2, PRO1598, RL
Chromosome Location:
Chr.7: 103471784 - 103989516 on Build GRCh38
UniGene ID:
Hs.655654
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS108624
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RELN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005045.3
NP_005036.2
44, 45
7084
NM_173054.2
NP_774959.1
44, 45
7084
GenBank mRNA
U79716.1
-
44, 45
7099
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix glycoprotein
extracellular matrix protein
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis involved in differentiation
neuron migration
proteolysis
cell adhesion
axon guidance
central nervous system development
brain development
long-term memory
associative learning
glial cell differentiation
positive regulation of neuron projection development
positive regulation of phosphatidylinositol 3-kinase signaling
dendrite development
peptidyl-tyrosine phosphorylation
spinal cord patterning
ventral spinal cord development
hippocampus development
cerebral cortex tangential migration
layer formation in cerebral cortex
positive regulation of TOR signaling
positive regulation of CREB transcription factor activity
reelin-mediated signaling pathway
positive regulation of protein kinase activity
response to pain
positive regulation of peptidyl-tyrosine phosphorylation
regulation of behavior
modulation of synaptic transmission
positive regulation of small GTPase mediated signal transduction
positive regulation of synaptic transmission, glutamatergic
long-term synaptic potentiation
positive regulation of dendritic spine morphogenesis
positive regulation of protein tyrosine kinase activity
positive regulation of synapse maturation
NMDA glutamate receptor clustering
postsynaptic density protein 95 clustering
receptor localization to synapse
lateral motor column neuron migration
positive regulation of long-term synaptic potentiation
positive regulation of lateral motor column neuron migration
regulation of N-methyl-D-aspartate selective glutamate receptor activity
positive regulation of excitatory postsynaptic potential
positive regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
protein serine/threonine/tyrosine kinase activity
serine-type peptidase activity
metal ion binding
lipoprotein particle receptor binding
very-low-density lipoprotein particle receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-77xhx:80/100.66.78.205:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0