Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270398.1 | NP_001257327.1 | 7 | 1223 | ||
NM_002821.4 | NP_002812.2 | 7 | 1350 | |||
NM_152880.3 | NP_690619.1 | 7 | 1350 | |||
NM_152881.3 | NP_690620.1 | 7 | 1350 | |||
NM_152882.3 | NP_690621.1 | 7 | 1350 | |||
NR_072997.1 | 6 | 1159 | ||||
NR_072998.1 | 7 | 1350 | ||||
XM_011514765.2 | XP_011513067.1 | 7 | 1218 | |||
XM_011514766.2 | XP_011513068.1 | 7 | 1218 | |||
GenBank mRNA | AB209350.1 | - | 7 | 1249 | ||
AF531868.1 | - | 7 | 1276 | |||
AF531869.1 | - | 7 | 1276 | |||
AF531870.1 | - | 7 | 1276 | |||
AF531871.1 | - | 7 | 1276 | |||
AF531872.1 | - | 7 | 1276 | |||
AK291016.1 | - | 7 | 1182 | |||
AK292589.1 | - | 7 | 1277 | |||
AK296931.1 | - | 8 | 1141 | |||
AK296953.1 | - | 7 | 1158 | |||
AL157486.1 | - | 1 | 63 | |||
BC046109.2 | - | 7 | 1290 | |||
BC071557.1 | - | 7 | 1304 | 30347652 | ||
U33635.1 | - | 7 | 1321 | |||
U40271.2 | - | 7 | 1276 |