Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193644.1 | NP_001180573.1 | 1, 2 | 94 | ||
NM_002857.3 | NP_002848.1 | 1, 2 | 94 | |||
NR_036493.1 | 1, 2 | 94 | ||||
GenBank mRNA | AB018541.1 | - | 1, 2 | 71 | ||
AK303099.1 | - | 1, 2 | 89 | |||
BC000496.2 | - | 1, 2 | 69 | 2820701 | ||
BT006879.1 | - | 1, 2 | 67 | |||
CB993363.1 | - | 111 | ||||
CD557913.1 | - | 1, 2 | 84 | |||
DC400252.1 | - | 1, 2 | 94 | |||
DC405033.1 | - | 1, 2 | 89 | |||
KU178290.1 | - | 1, 2 | 67 | |||
X75535.1 | - | 1, 2 | 77 |