Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014226.2 | NP_055041.1 | 3 | 422 | ||
XM_006720225.2 | XP_006720288.1 | 3 | 422 | |||
XM_006720226.2 | XP_006720289.1 | 3 | 422 | |||
XM_011537052.1 | XP_011535354.1 | 3 | 422 | |||
XM_017021549.1 | XP_016877038.1 | 3 | 422 | |||
XM_017021551.1 | XP_016877040.1 | 4 | 362 | |||
XM_017021555.1 | XP_016877044.1 | 3 | 422 | |||
XM_017021556.1 | XP_016877045.1 | 3 | 422 | |||
GenBank mRNA | AB022694.1 | - | 3 | 416 | ||
AK312778.1 | - | 3 | 413 | |||
BC026069.1 | - | 3 | 412 | 4838296 | ||
BC053536.1 | - | 3 | 398 | 5175851 | ||
DB037795.1 | - | 3 | 422 |