Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RANGAP1 SIRNA Assays ›
          Gene Symbol:
          RANGAP1
          Gene Name:
          Ran GTPase activating protein 1
          Gene Aliases:
          Fug1, RANGAP, SD
          Chromosome Location:
          Chr.22: 41244777 - 41302213 on Build GRCh38
          UniGene ID:
          Hs.183800
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS109045
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RANGAP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278651.1 NP_001265580.1 14, 15 2236
          NM_001317930.1 NP_001304859.1 13, 14 1690
          NM_002883.3 NP_002874.1 13, 14 1775
          XM_005261695.1 XP_005261752.1 13, 14 1903
          XM_005261696.1 XP_005261753.1 13, 14 1674
          XM_006724289.3 XP_006724352.1 13, 14 1678
          XM_011530293.1 XP_011528595.1 12, 13 1768
          XM_011530294.2 XP_011528596.1 13, 14 3384
          XM_011530295.2 XP_011528597.1 13, 14 2083
          XM_017028893.1 XP_016884382.1 14, 15 3014
          XM_017028894.1 XP_016884383.1 14, 15 3152
          XM_017028895.1 XP_016884384.1 13, 14 3514
          XM_017028896.1 XP_016884385.1 14, 15 1902
          XM_017028897.1 XP_016884386.1 13, 14 2994
          XM_017028898.1 XP_016884387.1 15, 16 2927
          XM_017028899.1 XP_016884388.1 12, 13 1640
          GenBank mRNA AB058738.1 - 12, 13 2959
          AK302222.1 - 12, 13 1339
          AK303524.1 - 13, 14 1698
          BC004990.1 - 3, 4 350
          BC014044.2 - 13, 14 1690 3506039
          BC041396.2 - 14, 15 2246 5277434
          CR456557.1 - 12, 13 1532
          X82260.1 - 12, 13 1471

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          sister chromatid cohesion
          signal transduction
          protein sumoylation
          positive regulation of GTPase activity
          negative regulation of protein export from nucleus
          response to axon injury
          cell-cell adhesion
          cellular response to vasopressin
          GTPase activator activity
          protein binding
          Ran GTPase binding
          ubiquitin protein ligase binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-n8jtk:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0