Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000537.3 | NP_000528.1 | 2, 3 | 300 | ||
GenBank mRNA | BC033474.1 | - | 2, 3 | 282 | 5188566 | |
BC047752.1 | - | 2, 3 | 285 | 5590211 | ||
BP321283.1 | - | 2, 3 | 300 | |||
CR536498.1 | - | 2, 3 | 256 |