Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256821.1 | NP_001243750.1 | 2 | 144 | ||
NM_006912.5 | NP_008843.1 | 2 | 266 | |||
GenBank mRNA | AF084462.1 | - | 2 | 177 | ||
AF493923.1 | - | 1 | 63 | |||
AK314239.1 | - | 2 | 242 | |||
BC104186.2 | - | 2 | 137 | 40030939 | ||
BC104187.2 | - | 2 | 137 | 40030940 | ||
BI916790.1 | - | 2 | 127 | |||
BM546985.1 | - | 2 | 260 | |||
BQ918446.1 | - | 2 | 144 | |||
CR407639.1 | - | 1 | 63 | |||
U71203.1 | - | 2 | 198 | |||
U78165.1 | - | 2 | 198 | |||
Y07566.1 | - | 2 | 208 |