Search
Search

SEC14L1| Gene Symbol: SEC14L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039573.2 | NP_001034662.2 | 4 | 404 | ||
| NM_001143998.1 | NP_001137470.1 | 4 | 404 | |||
| NM_001143999.1 | NP_001137471.1 | 4 | 400 | |||
| NM_001144001.1 | NP_001137473.1 | 2 | 192 | |||
| NM_001204408.1 | NP_001191337.1 | 6 | 749 | |||
| NM_001204410.1 | NP_001191339.1 | 5 | 442 | |||
| NM_003003.3 | NP_002994.3 | 4 | 351 | |||
| GenBank mRNA | AB208811.1 | - | 4 | 383 | ||
| AK290913.1 | - | 4 | 297 | |||
| AK293206.1 | - | 2 | 192 | |||
| BC136523.1 | - | 4 | 239 | 9020513 | ||
| BC136525.1 | - | 4 | 239 | 9020515 | ||
| BC142979.1 | - | 4 | 295 | 8860414 | ||
| BX537448.1 | - | 4 | 305 | |||
| D67029.1 | - | 4 | 387 | |||
| DA024354.1 | - | 4 | 342 | |||
| DB040556.1 | - | 4 | 404 | |||
| DB040580.1 | - | 2 | 192 | |||
| FM995495.1 | - | 6 | 643 | |||
| FM995496.1 | - | 5 | 440 | |||
| FM995497.1 | - | 5 | 539 | |||