Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003058.3 | NP_003049.2 | 4 | 998 | ||
GenBank mRNA | AB075951.1 | - | 4 | 1002 | ||
AK290787.1 | - | 4 | 902 | |||
BC030978.1 | - | 3 | 2161 | |||
BC039899.1 | - | 4 | 879 | 5186229 | ||
X98333.1 | - | 4 | 968 |