Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@3ad9429c
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Contact Us
Sign In
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
Gene Symbol:
SOX5
Gene Name:
SRY-box 5
Gene Aliases:
L-SOX5, L-SOX5B, L-SOX5F, LAMSHF
Chromosome Location:
Chr.12: 23529499 - 24562701 on Build GRCh38
UniGene ID:
Hs.657542
Species:
Human
siRNA ID
HSS110095
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price (USD)
401.00
Your Price (USD)
Online offer (USD):
401.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SOX5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001261415.1
NP_001248344.1
10
1380
NM_006940.4
NP_008871.3
10
1356
NM_152989.3
NP_694534.1
13
1589
NM_178010.2
NP_821078.1
2
156
XM_011520831.2
XP_011519133.1
10
1218
XM_011520832.2
XP_011519134.1
10
1330
XM_011520833.2
XP_011519135.1
10
1659
XM_011520834.2
XP_011519136.1
14
1792
XM_011520835.2
XP_011519137.1
13
1844
XM_011520837.2
XP_011519139.1
10
1401
XM_011520838.2
XP_011519140.1
10
1221
XM_011520842.2
XP_011519144.1
3
281
XM_017019888.1
XP_016875377.1
11
1673
XM_017019889.1
XP_016875378.1
11
1669
XM_017019890.1
XP_016875379.1
14
1808
XM_017019891.1
XP_016875380.1
14
1666
XM_017019892.1
XP_016875381.1
15
1887
XM_017019893.1
XP_016875382.1
12
1747
XM_017019894.1
XP_016875383.1
10
3677
XM_017019895.1
XP_016875384.1
10
1244
XM_017019896.1
XP_016875385.1
14
1805
XM_017019897.1
XP_016875386.1
14
1703
XM_017019898.1
XP_016875387.1
14
1700
XM_017019899.1
XP_016875388.1
13
1412
XM_017019900.1
XP_016875389.1
10
2026
XM_017019901.1
XP_016875390.1
10
6482
GenBank mRNA
AB081588.1
-
13
1587
AB081589.1
-
10
1356
AK074317.1
-
10
1283
AK096569.1
-
10
1650
AK098610.1
-
2
156
AK223536.1
-
2
156
AK299607.1
-
10
1268
AK303970.1
-
10
1380
BC029220.2
-
1
94
BC035227.2
-
1
94
BC047665.2
-
1
94
JX570584.1
-
11
1401
JX570585.1
-
11
1296
S83308.1
-
1
90
Back To Top
More Information
Panther Classification:
Molecular Function -
HMG box transcription factor
nucleic acid binding
transcription factor
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-3-prod-green:8080/10.253.229.154:8080.
git-commit: 0f6c802f6da5e59cd1b986a4dc444b4bcf3ac46d
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.15.0-2021.01.02-1.0