Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@466f2660
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Contact Us
Sign In
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
Gene Symbol:
SOX5
Gene Name:
SRY-box 5
Gene Aliases:
L-SOX5, L-SOX5B, L-SOX5F, LAMSHF
Chromosome Location:
Chr.12: 23529499 - 24562701 on Build GRCh38
UniGene ID:
Hs.657542
Species:
Human
siRNA ID
HSS110096
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SOX5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001261414.1
NP_001248343.1
14, 15
1693
NM_001261415.1
NP_001248344.1
12, 13
1733
NM_006940.4
NP_008871.3
12, 13
1709
NM_152989.3
NP_694534.1
15, 16
1942
NM_178010.2
NP_821078.1
4, 5
509
XM_011520831.2
XP_011519133.1
12, 13
1571
XM_011520832.2
XP_011519134.1
12, 13
1683
XM_011520833.2
XP_011519135.1
12, 13
2012
XM_011520834.2
XP_011519136.1
16, 17
2145
XM_011520835.2
XP_011519137.1
15, 16
2197
XM_011520837.2
XP_011519139.1
12, 13
1754
XM_011520838.2
XP_011519140.1
12, 13
1574
XM_011520842.2
XP_011519144.1
5, 6
634
XM_017019888.1
XP_016875377.1
13, 14
2026
XM_017019889.1
XP_016875378.1
13, 14
2022
XM_017019890.1
XP_016875379.1
16, 17
2161
XM_017019891.1
XP_016875380.1
16, 17
2019
XM_017019892.1
XP_016875381.1
17, 18
2240
XM_017019893.1
XP_016875382.1
14, 15
2100
XM_017019894.1
XP_016875383.1
12, 13
4030
XM_017019895.1
XP_016875384.1
12, 13
1597
XM_017019896.1
XP_016875385.1
16, 17
2158
XM_017019897.1
XP_016875386.1
16, 17
2056
XM_017019898.1
XP_016875387.1
16, 17
2053
XM_017019899.1
XP_016875388.1
15, 16
1765
XM_017019900.1
XP_016875389.1
12, 13
2379
XM_017019901.1
XP_016875390.1
12, 13
6835
GenBank mRNA
AB081588.1
-
15, 16
1940
AB081589.1
-
12, 13
1709
AK074317.1
-
12, 13
1636
AK096569.1
-
12, 13
2003
AK098610.1
-
4, 5
509
AK223536.1
-
4, 5
509
AK299607.1
-
11, 12
1512
AK303970.1
-
12, 13
1733
BC029220.2
-
3, 4
447
BC035227.2
-
3, 4
447
BC047665.2
-
3, 4
447
BC060773.1
-
14, 15
1694
30343519
CR456713.1
-
3, 4
359
JX570584.1
-
13, 14
1754
JX570585.1
-
13, 14
1649
S83308.1
-
3, 4
443
Back To Top
More Information
Panther Classification:
Molecular Function -
HMG box transcription factor
nucleic acid binding
transcription factor
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-3-prod-green:8080/10.253.229.154:8080.
git-commit: 0f6c802f6da5e59cd1b986a4dc444b4bcf3ac46d
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.15.0-2021.01.02-1.0