Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@2ba11eb2
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
See other TCF7L2 SIRNA Assays ›
Gene Symbol:
TCF7L2
Gene Name:
transcription factor 7 like 2
Gene Aliases:
TCF-4, TCF4
Chromosome Location:
Chr.10: 112950219 - 113167678 on Build GRCh38
UniGene ID:
Hs.593995
Species:
Human
siRNA ID
HSS110545
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price (USD)
401.00
Your Price (USD)
Online offer (USD):
401.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF7L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146274.1
NP_001139746.1
9
1490
NM_001146283.1
NP_001139755.1
9
1562
NM_001146284.1
NP_001139756.1
8
1409
NM_001146285.1
NP_001139757.1
8
1421
NM_001146286.1
NP_001139758.1
8
1421
NM_001198525.1
NP_001185454.1
8
1436
NM_001198526.1
NP_001185455.1
8
1421
NM_001198527.1
NP_001185456.1
8
1409
NM_001198528.1
NP_001185457.1
8
1421
NM_001198529.1
NP_001185458.1
8
1421
NM_001198530.1
NP_001185459.1
7
1319
NM_001198531.1
NP_001185460.1
9
1490
NM_030756.4
NP_110383.2
8
1421
XM_005270071.1
XP_005270128.1
10
1680
XM_005270075.1
XP_005270132.1
10
1665
XM_005270077.1
XP_005270134.1
10
1680
XM_005270078.1
XP_005270135.1
10
1680
XM_005270079.1
XP_005270136.1
10
1665
XM_005270080.1
XP_005270137.1
9
1611
XM_005270084.1
XP_005270141.1
9
1521
XM_005270085.1
XP_005270142.1
9
1536
XM_005270086.1
XP_005270143.1
8
1467
XM_005270088.1
XP_005270145.1
8
1440
XM_005270089.1
XP_005270146.1
8
1467
XM_005270091.2
XP_005270148.1
10
1680
XM_005270092.1
XP_005270149.1
10
1680
XM_005270093.2
XP_005270150.1
10
1680
XM_005270094.2
XP_005270151.1
10
1677
XM_005270095.1
XP_005270152.1
10
1680
XM_005270096.2
XP_005270153.1
9
1521
XM_005270100.1
XP_005270157.1
9
1536
XM_005270101.2
XP_005270158.1
8
1467
XM_005270102.1
XP_005270159.1
9
1509
XM_005270103.1
XP_005270160.1
8
1467
XM_005270104.1
XP_005270161.1
8
1440
XM_011540109.1
XP_011538411.1
10
1680
XM_011540110.1
XP_011538412.1
9
1584
XM_011540111.1
XP_011538413.1
8
1452
XM_011540113.2
XP_011538415.1
10
1680
XM_011540116.1
XP_011538418.1
10
1680
XM_017016584.1
XP_016872073.1
10
1677
XM_017016585.1
XP_016872074.1
10
1668
XM_017016586.1
XP_016872075.1
10
1665
XM_017016587.1
XP_016872076.1
10
1653
XM_017016588.1
XP_016872077.1
9
1611
XM_017016589.1
XP_016872078.1
10
1680
XM_017016590.1
XP_016872079.1
9
1536
XM_017016591.1
XP_016872080.1
9
1524
XM_017016592.1
XP_016872081.1
9
1521
XM_017016593.1
XP_016872082.1
9
1521
XM_017016594.1
XP_016872083.1
8
1467
XM_017016595.1
XP_016872084.1
10
1665
XM_017016596.1
XP_016872085.1
9
1509
GenBank mRNA
AB440195.1
-
9
983
AB451266.1
-
8
914
AK225809.1
-
8
1425
AK296305.1
-
5
786
AK299295.1
-
8
951
AK301483.1
-
6
710
AK303684.1
-
5
503
AK315243.1
-
9
1228
AK316111.1
-
5
499
BC032656.1
-
8
998
5533185
BX648364.1
-
9
858
CR536574.1
-
8
914
FJ010164.1
-
7
809
FJ010165.1
-
8
941
FJ010166.1
-
8
1022
FJ010167.1
-
8
1007
FJ010168.1
-
8
999
FJ010170.1
-
8
1007
FJ010171.1
-
7
849
FJ010172.1
-
8
1011
FJ010173.1
-
7
809
FJ010174.1
-
8
1369
HM352838.1
-
8
929
HM352839.1
-
8
929
HM352840.1
-
9
971
HM352841.1
-
9
998
HM352842.1
-
8
914
HM352843.1
-
8
902
HM352844.1
-
9
983
HM352845.1
-
9
1043
HM352846.1
-
9
983
HM352847.1
-
8
914
HM352848.1
-
8
914
HM352849.1
-
8
929
HM352850.1
-
9
983
HM352851.1
-
9
998
Y11306.2
-
8
1221
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleic acid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-2-prod-green:8080/10.253.226.220:8080.
git-commit: 0f6c802f6da5e59cd1b986a4dc444b4bcf3ac46d
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.15.0-2021.01.02-1.0