Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZEB1 SIRNA Assays ›
          Gene Symbol:
          ZEB1
          Gene Name:
          zinc finger E-box binding homeobox 1
          Gene Aliases:
          AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
          Chromosome Location:
          Chr.10: 31318495 - 31529814 on Build GRCh38
          UniGene ID:
          Hs.124503
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS110548
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZEB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001128128.2 NP_001121600.1 7 2049
          NM_001174093.1 NP_001167564.1 6 1709
          NM_001174094.1 NP_001167565.1 7 2046
          NM_001174095.1 NP_001167566.1 6 1568
          NM_001174096.1 NP_001167567.1 7 1772
          NM_001323638.1 NP_001310567.1 11 2419
          NM_001323641.1 NP_001310570.1 7 1999
          NM_001323642.1 NP_001310571.1 9 2301
          NM_001323643.1 NP_001310572.1 9 2079
          NM_001323644.1 NP_001310573.1 6 1798
          NM_001323645.1 NP_001310574.1 8 2142
          NM_001323646.1 NP_001310575.1 10 2380
          NM_001323647.1 NP_001310576.1 8 1912
          NM_001323648.1 NP_001310577.1 7 1852
          NM_001323649.1 NP_001310578.1 8 1915
          NM_001323650.1 NP_001310579.1 10 2147
          NM_001323651.1 NP_001310580.1 8 1815
          NM_001323652.1 NP_001310581.1 9 1964
          NM_001323653.1 NP_001310582.1 9 2071
          NM_001323654.1 NP_001310583.1 9 1958
          NM_001323655.1 NP_001310584.1 9 2068
          NM_001323656.1 NP_001310585.1 7 1743
          NM_001323657.1 NP_001310586.1 7 1746
          NM_001323658.1 NP_001310587.1 7 1585
          NM_001323659.1 NP_001310588.1 7 1803
          NM_001323660.1 NP_001310589.1 7 1800
          NM_001323661.1 NP_001310590.1 8 1946
          NM_001323662.1 NP_001310591.1 9 2348
          NM_001323663.1 NP_001310592.1 10 2287
          NM_001323664.1 NP_001310593.1 9 2271
          NM_001323665.1 NP_001310594.1 8 2189
          NM_001323666.1 NP_001310595.1 7 2129
          NM_001323671.1 NP_001310600.1 8 2025
          NM_001323672.1 NP_001310601.1 9 2077
          NM_001323673.1 NP_001310602.1 8 2022
          NM_001323674.1 NP_001310603.1 5 1547
          NM_001323675.1 NP_001310604.1 5 1657
          NM_001323676.1 NP_001310605.1 7 1882
          NM_001323677.1 NP_001310606.1 7 1879
          NM_001323678.1 NP_001310607.1 5 1824
          NM_030751.5 NP_110378.3 7 1769
          XM_006717498.2 XP_006717561.1 10 2207
          XM_011519643.2 XP_011517945.1 10 2510
          XM_017016597.1 XP_016872086.1 10 2109
          XM_017016598.1 XP_016872087.1 10 5239
          XM_017016599.1 XP_016872088.1 9 5077
          XM_017016600.1 XP_016872089.1 11 5318
          XM_017016601.1 XP_016872090.1 10 5236
          XM_017016602.1 XP_016872091.1 11 5315
          XM_017016603.1 XP_016872092.1 10 5153
          GenBank mRNA AK091478.1 - 6 1669
          AK294775.1 - 7 1964
          AK296244.1 - 7 1692
          AK299541.1 - 4 1306
          AK299666.1 - 7 1803
          AK300830.1 - 6 1516
          AK307510.1 - 8 1775
          AK314683.1 - 7 1730
          AL831979.1 - 9 2027
          BC009572.1 - 7 1725
          BC112392.1 - 7 1725 40036600
          BX647794.1 - 11 2420
          M81699.1 - 1 641
          U12170.1 - 7 1730

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          immune response
          pattern specification process
          central nervous system development
          cell proliferation
          negative regulation of cell proliferation
          regulation of mesenchymal cell proliferation
          regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of epithelial cell differentiation
          regulation of T cell differentiation in thymus
          positive regulation of neuron differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          embryonic camera-type eye morphogenesis
          embryonic skeletal system morphogenesis
          semicircular canal morphogenesis
          regulation of smooth muscle cell differentiation
          cartilage development
          cellular response to amino acid stimulus
          cochlea morphogenesis
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          transcription corepressor activity
          protein binding
          transcription factor binding
          zinc ion binding
          E-box binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0