Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173452.1 | NP_001166923.1 | 11 | 1812 | ||
NM_001173453.1 | NP_001166924.1 | 10 | 1659 | |||
NM_005653.4 | NP_005644.2 | 11 | 1812 | |||
GenBank mRNA | AK291264.1 | - | 11 | 1463 | ||
AK308087.1 | - | 11 | 1163 | |||
BC003634.2 | - | 11 | 1550 | 3529119 | ||
M84810.1 | - | 11 | 1454 | |||
U03494.1 | - | 11 | 1782 | |||
U03495.1 | - | 11 | 1446 |