Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TLE4 SIRNA Assays ›
          Gene Symbol:
          TLE4
          Gene Name:
          transducin like enhancer of split 4
          Gene Aliases:
          BCE-1, BCE1, E(spI), ESG, ESG4, GRG4, Grg-4
          Chromosome Location:
          Chr.9: 79571773 - 79726882 on Build GRCh38
          UniGene ID:
          Hs.444213
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS110795
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TLE4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282748.1 NP_001269677.1 14 2210
          NM_001282749.1 NP_001269678.1 12 2039
          NM_001282753.1 NP_001269682.1 11 1907
          NM_001282760.1 NP_001269689.1 14 2209
          NM_007005.4 NP_008936.2 13 2114
          NR_104239.1 14 2247
          XM_005252177.1 XP_005252234.1 13 1290
          XM_006717264.1 XP_006717327.1 16 1509
          XM_006717268.1 XP_006717331.1 15 1413
          XM_011518952.1 XP_011517254.1 16 1509
          XM_011518953.1 XP_011517255.1 16 1386
          XM_011518954.1 XP_011517256.1 15 1464
          XM_011518955.1 XP_011517257.1 15 1446
          XM_011518956.1 XP_011517258.1 15 1425
          XM_011518957.1 XP_011517259.1 15 1422
          XM_011518958.1 XP_011517260.1 15 1413
          XM_011518959.1 XP_011517261.1 14 1386
          XM_011518960.1 XP_011517262.1 14 1383
          XM_011518961.1 XP_011517263.1 13 1323
          XM_011518962.1 XP_011517264.1 13 1320
          XM_011518963.1 XP_011517265.1 13 1311
          XM_011518964.1 XP_011517266.1 13 1308
          XM_011518965.1 XP_011517267.1 13 1290
          XM_011518966.1 XP_011517268.1 13 1287
          XM_011518967.1 XP_011517269.1 14 1260
          XM_011518968.1 XP_011517270.1 12 1248
          XM_011518969.1 XP_011517271.1 13 1206
          XM_011518970.2 XP_011517272.1 16 1509
          XM_011518972.2 XP_011517274.1 4 336
          XM_017015067.1 XP_016870556.1 14 1314
          XM_017015068.1 XP_016870557.1 13 1341
          XM_017015069.1 XP_016870558.1 13 1169
          XM_017015070.1 XP_016870559.1 13 1246
          XM_017015071.1 XP_016870560.1 14 1326
          XM_017015072.1 XP_016870561.1 13 1247
          XM_017015073.1 XP_016870562.1 13 1249
          XM_017015074.1 XP_016870563.1 14 1260
          XM_017015075.1 XP_016870564.1 12 1242
          XM_017015076.1 XP_016870565.1 12 1189
          XM_017015077.1 XP_016870566.1 12 1227
          XM_017015078.1 XP_016870567.1 12 1224
          XM_017015079.1 XP_016870568.1 12 1152
          XM_017015080.1 XP_016870569.1 12 1215
          XM_017015081.1 XP_016870570.1 12 1137
          XM_017015082.1 XP_016870571.1 12 1107
          XM_017015083.1 XP_016870572.1 12 1134
          XM_017015084.1 XP_016870573.1 11 1059
          XM_017015085.1 XP_016870574.1 11 967
          XM_017015086.1 XP_016870575.1 10 1005
          GenBank mRNA AB033087.1 - 13 1175
          AK057236.1 - 14 1395
          AK296342.1 - 12 1068
          AK302709.1 - 13 1149
          AL162059.1 - 11 949
          BC036369.1 - 14 1409
          BC045650.1 - 11 1907 5268902
          BC059405.1 - 14 1368 5296117
          M99439.1 - 2 121

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          biological_process
          Wnt signaling pathway
          beta-catenin-TCF complex assembly
          molecular_function
          chromatin binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          transcription corepressor activity
          protein binding
          repressing transcription factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ksb6d:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0