Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TLR2 SIRNA Assays ›
          Gene Symbol:
          TLR2
          Gene Name:
          toll like receptor 2
          Gene Aliases:
          CD282, TIL4
          Chromosome Location:
          Chr.4: 153684080 - 153710643 on Build GRCh38
          UniGene ID:
          Hs.519033
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS110813
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TLR2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001318787.1 NP_001305716.1 4 789
          NM_001318789.1 NP_001305718.1 3 579
          NM_001318790.1 NP_001305719.1 3 556
          NM_001318791.1 NP_001305720.1 3 545
          NM_001318793.1 NP_001305722.1 3 541
          NM_001318795.1 NP_001305724.1 3 520
          NM_001318796.1 NP_001305725.1 3 505
          NM_003264.4 NP_003255.2 3 564
          XM_011532215.2 XP_011530517.1 3 489
          XM_011532216.2 XP_011530518.1 2 436
          XM_017008573.1 XP_016864062.1 4 1035
          XM_017008574.1 XP_016864063.1 4 717
          XM_017008575.1 XP_016864064.1 4 761
          XM_017008576.1 XP_016864065.1 2 1335
          GenBank mRNA AB445624.1 - 1 308
          AF051152.1 - 2 470
          AF502291.1 - 1 308
          AK125659.1 - 1 1712
          BC033756.1 - 3 458 5213439
          BI769109.1 - 3 527
          DA460298.1 - 4 530
          DA870971.1 - 4 545
          DQ012265.1 - 1 308
          DQ012266.1 - 1 308
          DQ012267.1 - 1 308
          DQ012268.1 - 1 308
          DQ012269.1 - 1 308
          DQ012270.1 - 1 308
          DQ012271.1 - 1 308
          KU178364.1 - 1 308
          U88878.1 - 2 437

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine secretion involved in immune response
          MyD88-dependent toll-like receptor signaling pathway
          apoptotic process
          inflammatory response
          immune response
          signal transduction
          I-kappaB phosphorylation
          positive regulation of gene expression
          positive regulation of Wnt signaling pathway
          lipopolysaccharide-mediated signaling pathway
          interleukin-10 production
          positive regulation of chemokine production
          positive regulation of interferon-beta production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-18 production
          positive regulation of interleukin-6 production
          positive regulation of interleukin-8 production
          positive regulation of tumor necrosis factor production
          positive regulation of toll-like receptor signaling pathway
          toll-like receptor 2 signaling pathway
          toll-like receptor TLR1:TLR2 signaling pathway
          toll-like receptor TLR6:TLR2 signaling pathway
          positive regulation of NF-kappaB import into nucleus
          detection of triacyl bacterial lipopeptide
          detection of diacyl bacterial lipopeptide
          innate immune response
          positive regulation of transcription from RNA polymerase II promoter
          regulation of cytokine secretion
          positive regulation of inflammatory response
          defense response to Gram-positive bacterium
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of nitric-oxide synthase biosynthetic process
          cellular response to bacterial lipopeptide
          cellular response to lipoteichoic acid
          cellular response to diacyl bacterial lipopeptide
          cellular response to triacyl bacterial lipopeptide
          lipopolysaccharide binding
          lipopolysaccharide receptor activity
          receptor activity
          transmembrane signaling receptor activity
          protein binding
          signaling pattern recognition receptor activity
          Toll-like receptor binding
          triacyl lipopeptide binding
          peptidoglycan binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0