Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNT3 SIRNA Assays ›
Gene Symbol:
TNNT3
Gene Name:
troponin T3, fast skeletal type
Gene Aliases:
TNTF
Chromosome Location:
Chr.11: 1919551 - 1938706 on Build GRCh38
UniGene ID:
Hs.73454
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS110884
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TNNT3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042780.2
NP_001036245.1
9
460
NM_001042781.2
NP_001036246.1
10
478
NM_001042782.2
NP_001036247.1
9
460
NM_001297646.1
NP_001284575.1
10
718
NM_006757.3
NP_006748.1
10
484
XM_006718288.3
XP_006718351.1
5
511
XM_006718290.3
XP_006718353.1
12
423
XM_006718293.2
XP_006718356.1
11
500
XM_006718294.3
XP_006718357.1
11
500
XM_006718296.3
XP_006718359.1
10
387
XM_006718299.2
XP_006718362.1
9
323
XM_006718300.3
XP_006718363.1
8
351
XM_006718302.3
XP_006718365.1
7
333
XM_011520343.2
XP_011518645.1
12
554
XM_017018205.1
XP_016873694.1
10
478
XM_017018206.1
XP_016873695.1
12
346
XM_017018207.1
XP_016873696.1
11
334
XM_017018208.1
XP_016873697.1
10
301
XM_017018209.1
XP_016873698.1
10
310
XM_017018210.1
XP_016873699.1
9
275
XM_017018211.1
XP_016873700.1
9
294
XM_017018212.1
XP_016873701.1
8
266
XM_017018213.1
XP_016873702.1
7
251
XM_017018214.1
XP_016873703.1
7
250
XM_017018215.1
XP_016873704.1
7
242
XM_017018216.1
XP_016873705.1
11
405
XM_017018217.1
XP_016873706.1
10
384
XM_017018218.1
XP_016873707.1
9
372
XM_017018219.1
XP_016873708.1
7
333
GenBank mRNA
AK056968.1
-
9
539
BC050446.1
-
9
316
BC117327.1
-
8
344
40125878
BC143537.1
-
8
344
9052047
BC171727.1
-
9
362
9052046
BC171728.1
-
5
297
BT019997.1
-
9
266
BX640689.1
-
10
342
CR541927.1
-
9
266
DA897990.1
-
10
478
DA899275.1
-
8
369
DQ778624.1
-
5
248
M21984.1
-
9
284
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
regulation of striated muscle contraction
muscle filament sliding
regulation of ATPase activity
actin binding
tropomyosin binding
troponin C binding
calcium-dependent ATPase activity
troponin I binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-x54js:80/100.66.79.231:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0