Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142407.2 | NP_001135879.1 | 9 | 838 | ||
NM_001142408.2 | NP_001135880.1 | 10 | 895 | |||
NM_001142417.2 | NP_001135889.1 | 10 | 875 | |||
NM_001142435.2 | NP_001135907.1 | 9 | 858 | |||
NM_001261822.1 | NP_001248751.1 | 9 | 864 | |||
NM_003296.3 | NP_003287.1 | 10 | 901 | |||
XM_005249349.1 | XP_005249406.1 | 11 | 980 | |||
XM_005249350.1 | XP_005249407.1 | 10 | 938 | |||
XM_005249351.3 | XP_005249408.1 | 10 | 958 | |||
XM_005249352.3 | XP_005249409.1 | 10 | 949 | |||
XM_005249353.3 | XP_005249410.1 | 11 | 985 | |||
XM_005249356.1 | XP_005249413.1 | 11 | 983 | |||
XM_011514841.1 | XP_011513143.1 | 11 | 1315 | |||
XM_011514842.1 | XP_011513144.1 | 10 | 1278 | |||
GenBank mRNA | AK128009.1 | - | 1 | 1831 | ||
AK223337.1 | - | 10 | 838 | |||
AK292385.1 | - | 10 | 879 | |||
AY289796.1 | - | 10 | 845 | |||
AY292862.1 | - | 10 | 940 | |||
AY292863.1 | - | 9 | 839 | |||
AY773479.1 | - | 10 | 881 | |||
BC022011.2 | - | 10 | 848 | 4826427 | ||
BC107707.1 | - | 9 | 795 | 6618679 | ||
BI831679.1 | - | 4 | 250 | |||
BM552843.1 | - | 9 | 773 | |||
BP369727.1 | - | 4 | 352 | |||
M25532.1 | - | 10 | 824 | |||
X95239.1 | - | 10 | 875 |