Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TSHR SIRNA Assays ›
          Gene Symbol:
          TSHR
          Gene Name:
          thyroid stimulating hormone receptor
          Gene Aliases:
          CHNG1, LGR3, hTSHR-I
          Chromosome Location:
          Chr.14: 80954989 - 81146302 on Build GRCh38
          UniGene ID:
          Hs.160411
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS111021
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TSHR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000369.2 NP_000360.2 5 595
          NM_001018036.2 NP_001018046.1 5 595
          NM_001142626.2 NP_001136098.1 5 595
          XM_005268037.4 XP_005268094.1 6 815
          XM_005268039.1 XP_005268096.1 6 815
          XM_006720245.1 XP_006720308.1 6 815
          XM_011537119.2 XP_011535421.1 5 540
          GenBank mRNA AB209207.1 - 5 467
          AY429111.1 - 5 439
          BC009237.2 - 5 504
          BC024205.2 - 5 689
          BC063613.1 - 5 471
          BC108653.1 - 5 512
          BC120972.2 - 5 465
          BC120973.2 - 5 465
          BC127628.1 - 5 465
          BC141970.1 - 5 465
          M31774.1 - 5 539
          M32215.1 - 5 483
          M73747.1 - 5 509
          S45272.1 - 5 439
          S49816.1 - 5 460

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          activation of adenylate cyclase activity
          cell-cell signaling
          nervous system development
          positive regulation of cell proliferation
          hormone-mediated signaling pathway
          thyroid-stimulating hormone signaling pathway
          thyroid-stimulating hormone receptor activity
          protein binding
          G-protein coupled peptide receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0