Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TNFSF4 SIRNA Assays ›
          Gene Symbol:
          TNFSF4
          Gene Name:
          tumor necrosis factor superfamily member 4
          Gene Aliases:
          CD134L, CD252, GP34, OX-40L, OX4OL, TNLG2B, TXGP1
          Chromosome Location:
          Chr.1: 173183729 - 173462208 on Build GRCh38
          UniGene ID:
          Hs.181097
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS111078
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TNFSF4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001297562.1 NP_001284491.1 3 607
          NM_003326.4 NP_003317.1 3 649
          XM_011509964.2 XP_011508266.2 4 1452
          XM_017002228.1 XP_016857717.1 3 1881
          XM_017002229.1 XP_016857718.1 5 6772
          XM_017002230.1 XP_016857719.1 4 1136
          GenBank mRNA AK297932.1 - 3 522
          BC041663.1 - 3 607 4510740
          CR542033.1 - 3 512
          D90224.1 - 3 548
          X79929.1 - 3 649

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          defense response to nematode
          acute inflammatory response
          positive regulation of T cell cytokine production
          regulation of adaptive immune response
          positive regulation of type 2 immune response
          positive regulation of immunoglobulin mediated immune response
          immune response
          signal transduction
          cholesterol metabolic process
          positive regulation of cell proliferation
          response to virus
          negative regulation of interferon-gamma production
          negative regulation of interleukin-17 production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-13 production
          positive regulation of interleukin-2 production
          positive regulation of interleukin-4 production
          positive regulation of interleukin-6 production
          tumor necrosis factor-mediated signaling pathway
          memory T cell activation
          T-helper 2 cell activation
          response to nitrogen dioxide
          cellular response to nitrogen dioxide
          CD4-positive, alpha-beta T cell costimulation
          T cell proliferation
          positive regulation of T cell proliferation
          positive regulation of activated T cell proliferation
          positive regulation of CD4-positive, alpha-beta T cell differentiation
          positive regulation of memory T cell differentiation
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of regulatory T cell differentiation
          negative regulation of T-helper 1 cell differentiation
          positive regulation of T-helper 2 cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of alpha-beta T cell proliferation
          negative regulation of cytokine secretion
          regulation of inflammatory response
          positive regulation of inflammatory response
          positive regulation of B cell activation
          positive regulation of immunoglobulin secretion
          cellular response to lipopolysaccharide
          cellular response to prostaglandin E stimulus
          chemokine (C-C motif) ligand 11 production
          positive regulation of CD4-positive, alpha-beta T cell costimulation
          positive regulation of memory T cell activation
          positive regulation of T-helper 2 cell activation
          positive regulation of interleukin-4-dependent isotype switching to IgE isotypes
          receptor binding
          cytokine activity
          tumor necrosis factor receptor binding
          tumor necrosis factor receptor superfamily binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0