Search
Search

ZNF35| Gene Symbol: ZNF35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003420.3 | NP_003411.3 | 4 | 1791 | ||
| XM_017007129.1 | XP_016862618.1 | 4 | 1765 | |||
| XM_017007130.1 | XP_016862619.1 | 4 | 1786 | |||
| XM_017007131.1 | XP_016862620.1 | 4 | 1787 | |||
| GenBank mRNA | AK300581.1 | - | 3 | 1645 | ||
| AK314821.1 | - | 4 | 1790 | |||
| BC013597.1 | - | 4 | 1769 | 4186299 | ||
| BC015892.1 | - | 3 | 1623 | |||
| X07289.1 | - | 5 | 1675 | |||