Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF113A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006978.2 | NP_008909.1 | 1 | 1139 | ||
GenBank mRNA | AF086291.1 | - | 1 | 431 | ||
AK314778.1 | - | 1 | 1085 | |||
BC000832.1 | - | 1 | 982 | 3455861 | ||
BC020556.1 | - | 1 | 1025 | 3447759 | ||
BT007175.1 | - | 1 | 924 | |||
X98253.1 | - | 1 | 1134 |