Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF195 SIRNA Assays ›
Gene Symbol:
ZNF195
Gene Name:
zinc finger protein 195
Gene Aliases:
HRF1, ZNFP104
Chromosome Location:
Chr.11: 3357927 - 3379222 on Build GRCh38
UniGene ID:
Hs.386294
Species:
Human
siRNA ID
HSS111667
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF195
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130519.2
NP_001123991.1
5
1296
NM_001130520.2
NP_001123992.1
6
1365
NM_001242841.1
NP_001229770.1
6
1376
NM_001242842.1
NP_001229771.1
6
1298
NM_001242843.1
NP_001229772.1
5
1229
NM_001256823.1
NP_001243752.1
6
1286
NM_001256824.1
NP_001243753.1
6
1322
NM_001256825.1
NP_001243754.1
5
1248
NM_007152.4
NP_009083.2
4
1149
NR_040083.1
6
1513
NR_046381.1
7
1433
NR_046382.1
5
1335
XM_006718306.1
XP_006718369.1
5
1352
XM_011520350.2
XP_011518652.1
7
1354
XM_011520351.2
XP_011518653.1
8
1725
XM_011520352.2
XP_011518654.1
6
1421
XM_017018260.1
XP_016873749.1
7
1568
XM_017018261.1
XP_016873750.1
7
1450
XM_017018262.1
XP_016873751.1
6
1499
XM_017018263.1
XP_016873752.1
6
1314
XM_017018264.1
XP_016873753.1
4
1166
XM_017018265.1
XP_016873754.1
7
1429
XM_017018266.1
XP_016873755.1
6
1349
XM_017018267.1
XP_016873756.1
8
1702
GenBank mRNA
AB209664.1
-
1
9282
AF003540.1
-
6
1228
AK095720.1
-
4
1072
AK290099.1
-
6
1274
AK293679.1
-
6
1298
AK308024.1
-
6
1309
AK308209.1
-
5
1189
AL833722.1
-
4
1062
BC121028.1
-
4
1005
40117443
BC121029.1
-
5
1152
40117445
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-5svh6:80/10.239.165.170:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0