Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SORBS2 SIRNA Assays ›
          Gene Symbol:
          SORBS2
          Gene Name:
          sorbin and SH3 domain containing 2
          Gene Aliases:
          ARGBP2, PRO0618
          Chromosome Location:
          Chr.4: 185585444 - 185956716 on Build GRCh38
          UniGene ID:
          Hs.481342
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS112345
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SORBS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145670.1 NP_001139142.1 8, 9 1330
          NM_001145671.2 NP_001139143.1 7, 8 858
          NM_001145672.1 NP_001139144.1 6, 7 655
          NM_001145673.1 NP_001139145.1 7, 8 1382
          NM_001270771.1 NP_001257700.1 9, 10 1339
          NM_003603.6 NP_003594.3 7, 8 1172
          NM_021069.4 NP_066547.1 6, 7 1119
          XM_005263307.1 XP_005263364.1 9, 10 1976
          XM_005263308.1 XP_005263365.1 8, 9 1334
          XM_005263310.1 XP_005263367.1 8, 9 1265
          XM_005263311.1 XP_005263368.1 8, 9 1334
          XM_005263312.1 XP_005263369.1 7, 8 1172
          XM_005263313.1 XP_005263370.1 8, 9 1334
          XM_006714388.1 XP_006714451.1 8, 9 1265
          XM_006714390.1 XP_006714453.1 7, 8 1241
          XM_017008738.1 XP_016864227.1 10, 11 2450
          XM_017008739.1 XP_016864228.1 11, 12 2753
          XM_017008740.1 XP_016864229.1 10, 11 2446
          XM_017008741.1 XP_016864230.1 10, 11 2450
          XM_017008742.1 XP_016864231.1 9, 10 2400
          XM_017008743.1 XP_016864232.1 10, 11 2381
          XM_017008744.1 XP_016864233.1 10, 11 2660
          XM_017008745.1 XP_016864234.1 9, 10 2600
          XM_017008746.1 XP_016864235.1 10, 11 2450
          XM_017008747.1 XP_016864236.1 10, 11 2591
          XM_017008748.1 XP_016864237.1 10, 11 2450
          XM_017008749.1 XP_016864238.1 10, 11 2012
          XM_017008750.1 XP_016864239.1 10, 11 2450
          XM_017008751.1 XP_016864240.1 10, 11 2450
          XM_017008752.1 XP_016864241.1 10, 11 2450
          XM_017008753.1 XP_016864242.1 10, 11 2450
          XM_017008754.1 XP_016864243.1 10, 11 2450
          XM_017008755.1 XP_016864244.1 7, 8 1415
          XM_017008756.1 XP_016864245.1 6, 7 1118
          XM_017008758.1 XP_016864247.1 7, 8 1284
          XM_017008759.1 XP_016864248.1 7, 8 2011
          XM_017008760.1 XP_016864249.1 7, 8 1241
          XM_017008761.1 XP_016864250.1 10, 11 2450
          XM_017008762.1 XP_016864251.1 8, 9 1334
          XM_017008763.1 XP_016864252.1 7, 8 1284
          XM_017008764.1 XP_016864253.1 8, 9 1265
          XM_017008765.1 XP_016864254.1 9, 10 1568
          XM_017008766.1 XP_016864255.1 7, 8 1241
          XM_017008767.1 XP_016864256.1 8, 9 1544
          XM_017008768.1 XP_016864257.1 7, 8 1462
          XM_017008769.1 XP_016864258.1 8, 9 1265
          XM_017008770.1 XP_016864259.1 7, 8 1241
          XM_017008771.1 XP_016864260.1 6, 7 1122
          XM_017008772.1 XP_016864261.1 7, 8 1172
          XM_017008773.1 XP_016864262.1 8, 9 1475
          XM_017008775.1 XP_016864264.1 10, 11 2450
          GenBank mRNA AB018320.1 - 6, 7 771
          AF049884.1 - 7, 8 1136
          AF049885.1 - 6, 7 501
          AK056628.1 - 9, 10 957
          AK225327.1 - 8, 9 1296
          AK225812.1 - 7, 8 1382
          AK293400.1 - 6, 7 655
          AK295742.1 - 7, 8 690
          BC011883.1 - 7, 8 825 3677070

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          actin filament organization
          cell adhesion
          biological_process
          cell migration
          cell growth involved in cardiac muscle cell development
          structural constituent of cytoskeleton
          protein binding
          cytoskeletal adaptor activity
          structural constituent of muscle
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gd6qx:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0