Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@2420131f
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
Gene Symbol:
PPFIA1
Gene Name:
PTPRF interacting protein alpha 1
Gene Aliases:
LIP.1, LIP1, LIPRIN
Chromosome Location:
Chr.11: 70270687 - 70384501 on Build GRCh38
UniGene ID:
Hs.530749
Species:
Human
siRNA ID
HSS112397
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003626.3
NP_003617.1
25, 26
3587
NM_177423.2
NP_803172.1
25, 26
3587
NR_045286.1
26, 27
3598
XM_006718716.3
XP_006718779.1
30, 31
3912
XM_011545306.2
XP_011543608.1
30, 31
3784
XM_011545307.2
XP_011543609.1
29, 30
3885
XM_011545308.2
XP_011543610.1
29, 30
3882
XM_011545309.2
XP_011543611.1
30, 31
3912
XM_011545310.2
XP_011543612.1
29, 30
3837
XM_011545311.2
XP_011543613.1
28, 29
3825
XM_011545312.2
XP_011543614.1
28, 29
3822
XM_011545313.2
XP_011543615.1
29, 30
3795
XM_011545314.2
XP_011543616.1
28, 29
3765
XM_011545315.2
XP_011543617.1
28, 29
3720
XM_011545316.2
XP_011543618.1
27, 28
3705
XM_011545317.2
XP_011543619.1
26, 27
3675
XM_011545318.2
XP_011543620.1
15, 16
2017
XM_011545319.1
XP_011543621.1
27, 28
3693
XM_017018448.1
XP_016873937.1
27, 28
3792
XM_017018449.1
XP_016873938.1
26, 27
3630
GenBank mRNA
AB209314.1
-
26, 27
3525
AK123331.1
-
2164
AK124621.1
-
5, 6
2132
BC034046.1
-
25, 26
3550
4794300
BX648037.1
-
24, 25
2887
U22815.1
-
25, 26
3601
U22816.1
-
25, 26
3601
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-2-prod-blue:8080/10.253.227.150:8080.
git-commit: 8ac494bcbcad7ea42f0639902f1f4fe0b41a6635
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.20.0-2021.04.15-1.0