Search
Search

LGR5| Gene Symbol: LGR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277226.1 | NP_001264155.1 | 7 | 1075 | ||
| NM_001277227.1 | NP_001264156.1 | 6 | 859 | |||
| NM_003667.3 | NP_003658.1 | 7 | 1075 | |||
| NR_110596.1 | 7 | 1075 | ||||
| GenBank mRNA | AB105190.1 | - | 7 | 1044 | ||
| AF061444.1 | - | 7 | 764 | |||
| AF062006.1 | - | 7 | 812 | |||
| AK075399.1 | - | 7 | 861 | |||
| AK294985.1 | - | 7 | 1076 | |||
| AK315133.1 | - | 7 | 1052 | |||
| BC096324.3 | - | 7 | 810 | 40008253 | ||
| BC096325.1 | - | 7 | 810 | 40008255 | ||
| BC096326.3 | - | 7 | 810 | 40008257 | ||
| BC099650.1 | - | 7 | 810 | 40008256 | ||
| FN820440.1 | - | 6 | 596 | |||