Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MADD SIRNA Assays ›
Gene Symbol:
MADD
Gene Name:
MAP kinase activating death domain
Gene Aliases:
DENN, IG20, RAB3GEP
Chromosome Location:
Chr.11: 47269376 - 47330031 on Build GRCh38
UniGene ID:
Hs.82548
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS112548
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MADD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135943.1
NP_001129415.1
27
4221
NM_001135944.1
NP_001129416.1
27
4188
NM_003682.3
NP_003673.3
30
4506
NM_130470.2
NP_569826.2
27
4495
NM_130471.2
NP_569827.2
28
4260
NM_130472.2
NP_569828.2
27
4200
NM_130473.2
NP_569829.2
28
4389
NM_130474.2
NP_569830.2
27
4200
NM_130475.2
NP_569831.1
30
4530
NM_130476.2
NP_569832.2
27
4350
XM_005253189.2
XP_005253246.1
30
4530
XM_005253196.2
XP_005253253.1
28
4413
XM_005253199.2
XP_005253256.1
27
4353
XM_005253200.2
XP_005253257.1
30
4527
XM_005253201.2
XP_005253258.1
28
4284
XM_005253203.2
XP_005253260.1
27
4224
XM_005253204.2
XP_005253261.1
27
4212
XM_005253205.1
XP_005253262.1
27
4224
XM_017018478.1
XP_016873967.1
30
4518
XM_017018479.1
XP_016873968.1
29
4464
XM_017018480.1
XP_016873969.1
28
4410
XM_017018481.1
XP_016873970.1
28
4404
XM_017018482.1
XP_016873971.1
28
4401
XM_017018483.1
XP_016873972.1
30
4389
XM_017018484.1
XP_016873973.1
27
4341
XM_017018485.1
XP_016873974.1
27
4302
XM_017018486.1
XP_016873975.1
29
4335
XM_017018487.1
XP_016873976.1
29
4329
XM_017018488.1
XP_016873977.1
30
4518
XM_017018489.1
XP_016873978.1
30
4479
XM_017018490.1
XP_016873979.1
29
4463
XM_017018491.1
XP_016873980.1
29
4429
XM_017018492.1
XP_016873981.1
29
4464
XM_017018493.1
XP_016873982.1
29
4445
XM_017018494.1
XP_016873983.1
28
4416
XM_017018495.1
XP_016873984.1
28
4413
XM_017018496.1
XP_016873985.1
28
4360
XM_017018497.1
XP_016873986.1
28
4365
XM_017018498.1
XP_016873987.1
30
4389
XM_017018499.1
XP_016873988.1
30
4358
XM_017018500.1
XP_016873989.1
27
4350
XM_017018501.1
XP_016873990.1
29
4303
XM_017018502.1
XP_016873991.1
29
4316
XM_017018503.1
XP_016873992.1
29
4300
XM_017018504.1
XP_016873993.1
29
4313
XM_017018505.1
XP_016873994.1
29
4291
XM_017018506.1
XP_016873995.1
29
4296
XM_017018507.1
XP_016873996.1
29
4316
XM_017018508.1
XP_016873997.1
28
4287
XM_017018509.1
XP_016873998.1
28
4240
XM_017018510.1
XP_016873999.1
28
4275
GenBank mRNA
AB002356.1
-
30
4502
AF440100.1
-
30
4480
AF440101.1
-
28
4363
AF440102.1
-
28
4234
AF440103.1
-
27
4174
AF440434.1
-
27
4174
AK292442.1
-
27
4152
AK308780.1
-
11
958
BC040484.1
-
27
4205
5271798
U44953.1
-
27
4314
U77352.1
-
27
4463
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
cell surface receptor signaling pathway
regulation of tumor necrosis factor-mediated signaling pathway
regulation of Rab protein signal transduction
regulation of apoptotic process
positive regulation of GTPase activity
regulation of cell cycle
execution phase of apoptosis
regulation of extrinsic apoptotic signaling pathway via death domain receptors
regulation of extrinsic apoptotic signaling pathway
death receptor binding
protein binding
Rab guanyl-nucleotide exchange factor activity
protein kinase activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-pzbd9:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0