Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CADPS SIRNA Assays ›
Gene Symbol:
CADPS
Gene Name:
calcium dependent secretion activator
Gene Aliases:
CADPS1, CAPS, CAPS1, UNC-31
Chromosome Location:
Chr.3: 62398346 - 62875389 on Build GRCh38
UniGene ID:
Hs.654933
Species:
Human
siRNA ID
HSS112616
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CADPS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003716.3
NP_003707.2
11
2189
NM_183393.2
NP_899630.1
11
2189
NM_183394.2
NP_899631.1
11
2189
XM_006713378.2
XP_006713441.2
11
2184
XM_011534177.1
XP_011532479.1
11
2184
XM_011534178.1
XP_011532480.1
11
2184
XM_011534180.1
XP_011532482.1
11
2184
XM_011534182.1
XP_011532484.1
11
2184
XM_011534183.1
XP_011532485.1
11
2184
XM_011534189.1
XP_011532491.1
11
2184
XM_011534191.2
XP_011532493.1
11
1829
XM_011534192.1
XP_011532494.1
11
2184
XM_011534193.1
XP_011532495.1
11
2184
XM_011534194.2
XP_011532496.1
11
1829
XM_011534195.1
XP_011532497.1
11
2184
XM_011534196.1
XP_011532498.1
11
2184
XM_011534197.2
XP_011532499.1
11
2142
XM_011534198.2
XP_011532500.1
11
1829
XM_011534199.2
XP_011532501.1
11
1829
XM_011534200.2
XP_011532502.1
11
1829
XM_011534202.2
XP_011532504.1
11
2184
XM_011534203.1
XP_011532505.1
11
2184
XM_017007358.1
XP_016862847.1
11
2184
XM_017007359.1
XP_016862848.1
11
2184
XM_017007360.1
XP_016862849.1
11
2184
XM_017007361.1
XP_016862850.1
11
2184
XM_017007362.1
XP_016862851.1
11
2184
XM_017007363.1
XP_016862852.1
11
2184
XM_017007364.1
XP_016862853.1
11
2184
XM_017007365.1
XP_016862854.1
11
2184
XM_017007366.1
XP_016862855.1
11
2184
XM_017007367.1
XP_016862856.1
11
1829
XM_017007368.1
XP_016862857.1
11
1829
XM_017007369.1
XP_016862858.1
11
2184
XM_017007370.1
XP_016862859.1
11
2184
XM_017007371.1
XP_016862860.1
11
2184
XM_017007372.1
XP_016862861.1
11
1829
XM_017007373.1
XP_016862862.1
11
2184
XM_017007374.1
XP_016862863.1
11
2184
XM_017007375.1
XP_016862864.1
11
1829
XM_017007376.1
XP_016862865.1
11
2184
XM_017007377.1
XP_016862866.1
11
1829
XM_017007378.1
XP_016862867.1
11
1829
XM_017007379.1
XP_016862868.1
11
2184
XM_017007380.1
XP_016862869.1
11
2184
XM_017007381.1
XP_016862870.1
11
2184
XM_017007382.1
XP_016862871.1
11
1829
XM_017007383.1
XP_016862872.1
11
2184
XM_017007384.1
XP_016862873.1
11
2184
XM_017007385.1
XP_016862874.1
11
2184
XM_017007386.1
XP_016862875.1
11
2184
GenBank mRNA
AB032947.2
-
11
2179
AB593079.1
-
11
2142
AF458662.1
-
11
2139
AL833895.1
-
11
1486
BC131731.1
-
4
315
40124561
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
protein transport
vesicle organization
synaptic vesicle priming
catecholamine secretion
dense core granule exocytosis
protein binding
lipid binding
protein kinase binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-zbtnl:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0