Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE8B SIRNA Assays ›
Gene Symbol:
PDE8B
Gene Name:
phosphodiesterase 8B
Gene Aliases:
ADSD, PPNAD3
Chromosome Location:
Chr.5: 77180252 - 77428256 on Build GRCh38
UniGene ID:
Hs.584830
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS112623
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE8B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001029851.2
NP_001025022.1
14, 15
1668
NM_001029852.2
NP_001025023.1
16, 17
1794
NM_001029853.2
NP_001025024.1
16, 17
1899
NM_001029854.2
NP_001025025.1
16, 17
1818
NM_003719.3
NP_003710.1
17, 18
1959
XM_005248621.4
XP_005248678.1
17, 18
2185
XM_005248623.4
XP_005248680.1
17, 18
1810
XM_005248624.4
XP_005248681.1
17, 18
1754
XM_006714725.3
XP_006714788.1
18, 19
2252
XM_006714726.3
XP_006714789.1
18, 19
2249
XM_011543699.2
XP_011542001.1
18, 19
2254
XM_011543700.2
XP_011542002.1
15, 16
1962
XM_011543701.2
XP_011542003.1
18, 19
1765
XM_011543702.2
XP_011542004.1
17, 18
1596
XM_011543703.2
XP_011542005.1
17, 18
1614
XM_011543704.2
XP_011542006.1
15, 16
1417
XM_017010003.1
XP_016865492.1
17, 18
2187
XM_017010004.1
XP_016865493.1
17, 18
1593
XM_017010005.1
XP_016865494.1
14, 15
1895
XM_017010006.1
XP_016865495.1
18, 19
2032
XM_017010007.1
XP_016865496.1
17, 18
1800
XM_017010008.1
XP_016865497.1
14, 15
1509
GenBank mRNA
AB085824.1
-
17, 18
1923
AB085825.1
-
14, 15
1629
AB085826.1
-
16, 17
1755
AB085827.1
-
16, 17
1860
AF079529.2
-
17, 18
1959
AK002074.1
-
6, 7
709
AK023913.1
-
12, 13
1161
AL831924.1
-
17, 18
1789
AY129948.1
-
16, 17
1818
AY129949.1
-
14, 15
1668
AY423729.1
-
2, 3
319
BC035144.1
-
4, 5
415
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
cAMP catabolic process
signal transduction
cyclic nucleotide metabolic process
negative regulation of insulin secretion
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline