Search
Search

DNAH17| Gene Symbol: DNAH17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173628.3 | NP_775899.3 | 69 | 11257 | ||
| XM_011525416.2 | XP_011523718.1 | 69 | 11293 | |||
| XM_017025261.1 | XP_016880750.1 | 27 | 4517 | |||
| GenBank mRNA | AB290170.1 | - | 12 | 1958 | ||
| AJ000522.1 | - | 8 | 1284 | |||
| AK126059.1 | - | 1 | 794 | |||
| AK128065.1 | - | 16 | 2794 | |||
| AL122077.1 | - | 10 | 1580 | |||
| AL832652.1 | - | 14 | 2328 | |||